Skip to Content
MilliporeSigma
All Photos(1)

Documents

HG000C41

Millipore

Baird-Parker Agar

powder, pH 6.8±0.2 (25 °C, after autoclaving), poly bottle of 12 × 500 g

Synonym(s):

Baird-Parker Agar Base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111

form

powder

composition

Agar, 20.0 g/L
Glycine, 12.0 g/L
Lithium Chloride, 5.0 g/L
Meat Extract, 5.0 g/L
Sodium Pyruvate, 10.0 g/L
Tryptone, 10.0 g/L
Yeast Extract, 1.0 g/L

packaging

poly bottle of 12 × 500 g

pH

6.8±0.2 (25 °C, after autoclaving)

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Application

A selective medium for the isolation and enumeration of coagulase-positive staphylococci primarily from foodstuffs.

Preparation Note

Suspend 63g in 1 litre demineralised water. Boil whilst stirring until completely dissolved. Autoclave at 121°C for 15 minutes. Cool to 45 - 50°C and aseptically add 1% of 1% sterile Potassium Tellurite Solution (HG00BX15.PK) and 5% sterile Egg Yolk Emulsion (HG00BX14.100). Homogenate and pour plates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service