Skip to Content
MilliporeSigma
All Photos(1)

Documents

75805

Millipore

Oxford Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Listeria Selective Agar, Oxford, Listeria Oxford Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 10 g/L
corn starch, 1 g/L
esculin, 1 g/L
ferric ammonium citrate, 0.5 g/L
lithium chloride, 15 g/L
sodium chloride, 5 g/L
special peptone, 23 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages
veterinary

microbiology

suitability

selective and differential for Listeria spp.

Application

For the isolation and cultivation of Listeria monocytogenes from specimens containing a mixed bacterial flora.

Preparation Note

Dissolve 27.8 g in 500 ml distilled water and mix thoroughly. Bring to the boil by gently heating. Autoclave at 121°C for 15 minutes and cool to 45-50°C. Aseptically add 1 vial of sterile supplement (Cat. No. 1.07006.0010). Mix well and distribute.

Other Notes

Selective differential medium for the isolation of L. monocytogenes

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Curtis, G.D.W., et al.
Letters in Applied Microbiology, 8, 95-95 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service