Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HATF08130

Millipore

Immobilon® -NC Nitrocellulose Membrane

20 sheets, 8.5 cm x 13.5 cm, 0.45 µm pore size, Triton-free, nitrocellulose-based transfer membrane

Synonym(s):

Western blotting membrane, blotting membrane, nitrocellulose transfer membrane, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

Product Name

Immobilon®-NC Transfer Membrane Midi Sheets, 20 sheets, 8.5 cm x 13.5 cm, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane

material

mixed cellulose esters (MCE) membrane
nitrocellulose membrane
plain filter
white filter

Quality Level

feature

hydrophilic

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

filter L × W

8.5 cm × 13.5 cm

pore size

0.45 μm pore size

capacity

259 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with India ink
for use with Ponceau¬S red

detection method

colorimetric
fluorometric
radioactive

shipped in

ambient

General description

Immobilon®-NC (HATF) membrane is composed of a mixed cellulose esters matrix.

Application

Immobilon®-NC Transfer Membrane has been used in western blot analysis.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service