Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF342

Sigma-Aldrich

LIF,
Human Recombinant Animal Free

Synonym(s):

Leukemia Inhibitory Factor, Emfilermin, MLPLI, HILDA, D-FACTOR

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$664.00

$664.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$664.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

$664.00


Please contact Customer Service for Availability

Request a Bulk Order

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Level

UniProt accession no.

shipped in

dry ice

General description

LIF is a pleiotrophic factor produced by multiple cell types including T cells, myelomoncytic lineages, fibroblasts, liver, heart and melanoma. LIF promotes long-term maintenance of embryonic stem cells by suppressing spontaneous differentiation. Other activities include the stimulation of acute phase protein synthesis by hepatocytes, stimulation of differentiation of cholinergic nerves, and suppression of adipogenesis by inhibiting the lipoprotein lipase in adipocytes. While human LIF is active on mouse cells and is widely used in the maintenance of murine ESC to prevent spontaneous differentiation, mouse LIF is not active on human cells due to its inability to bind to the human LIF receptor.
Recombinant human LIF is a 19.6 kDa protein containing 180 amino acids residues including three disulfide bonds.

Quality

Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 is ≤0.1 ng/ml, corresponding to a specific activity of ≥1 x 10^7 units/mg.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lizhao Feng et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(23), 2002155-2002155 (2020-12-12)
Canavan disease (CD) is a fatal leukodystrophy caused by mutation of the aspartoacylase (ASPA) gene, which leads to deficiency in ASPA activity, accumulation of the substrate N-acetyl-L-aspartate (NAA), demyelination, and spongy degeneration of the brain. There is neither a cure

Articles

Leukemia Inhibitory Factor (LIF) is a stem cell growth factor used for the in vitro culture of pluripotent mouse embryonic stem cells (ES cells).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service