Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DX1545

Supelco

N,N-Dimethylacetamide

Synonym(s):

N,N-Dimethylacetamide, DMAC

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
$776.00
200 L
$10,520.00

$776.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
4 L
$776.00
200 L
$10,520.00

About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12352111
EC Index Number:
204-826-4
bp:
165-166 °C/1013 hPa
vapor pressure:
1.76 hPa ( 20 °C)

$776.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1.76 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

color

APHA: ≤30

pH

4 (20 °C, 200 g/L in H2O)

bp

165-166 °C/1013 hPa

mp

-20 °C

transition temp

flash point 64 °C

solubility

soluble >1000 g/L

density

0.94 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

N(C)(C)C(=O)C

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Application


  • Ternary Phase-Field Simulation of Poly(vinylidene fluoride) Microporous Membrane Structures Prepared by Nonsolvent-Induced Phase Separation with Different Additives and Solvent Treatments: This study highlights the use of N,N-Dimethylacetamide (DMAC) in the preparation of advanced poly(vinylidene fluoride) membranes, showcasing its critical role in enhancing polymer processing and membrane structures in material science (Zhang et al., 2024).

  • Improved Protein Removal Performance of PES Hollow-Fiber Ultrafiltration Membrane with Sponge-like Structure: Research demonstrates DMAC′s effectiveness in the production of high-performance ultrafiltration membranes, emphasizing its importance in biomedical applications and wastewater treatment (Zhao et al., 2024).

  • Poly(vinylidene fluoride) Aerogels with alpha, beta and gamma; Crystalline Forms: Correlating Physicochemical Properties with Polymorphic Structures: This article showcases the versatility of DMAC as a solvent in the synthesis of poly(vinylidene fluoride) aerogels, linking its use to significant advancements in the understanding of polymer crystallinity and properties (Suresh et al., 2024).

  • Cellulose Esters: Synthesis for Further Formation of Films with Magnetite Nanoparticles Incorporated: The study leverages DMAC for the synthesis of advanced cellulose ester films integrated with magnetite nanoparticles, illustrating its potential in developing multifunctional materials for diverse industrial applications (Furlan Sandrini et al., 2024).

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Assay (GC): 98% min
Color (APHA): 30 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service