Skip to Content
MilliporeSigma
All Photos(1)

Documents

DX1543

Supelco

N,N-Dimethylacetamide

OmniSolv®, ≥99.9% (GC)

Synonym(s):

N,N-Dimethylacetamide, DMAC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12352111
EC Index Number:
204-826-4

vapor pressure

1.76 hPa ( 20 °C)

Quality Level

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.05% Water (H2O)

evapn. residue

≤5 ppm

color

APHA: ≤10

pH

4 (20 °C, 200 g/L in H2O)

bp

165-166 °C/1013 hPa

mp

-20 °C

transition temp

flash point 64 °C

density

0.931-0.941  g/mL at 25 °C

UV cutoff

≤270 nm

UV absorption

λ: 270 nm Amax: ≤1.00
λ: 280 nm Amax: ≤0.30
λ: 290 nm Amax: ≤0.15
λ: 310 nm Amax: ≤0.05
λ: 360 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Related Categories

Application

OmniSolv®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minColor (APHA)10 maxDensity at 25°C0.931-0.941g/mLEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardTitrable acidµeq/gTitrable baseµeq/gUV Abs. at 270 nm1.00AU maxUV Abs. at 280 nm0.30AU maxUV Abs. at 290 nm0.15AU maxUV Abs. at 310 nm0.05AU maxUV Abs. at 360 nm0.01AU maxUV Cut-off270nm maxWater (H2O)0.05% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service