Skip to Content
MilliporeSigma
All Photos(1)

Documents

DR1025

Sigma-Aldrich

PhosphoDetect Anti-Chk1 (pSer³¹⁷) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

monkey, rat, mouse, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer317/pSer317)

Gene Information

human ... CHEK1(1111)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~56 kDa Chk1 protein when phosphorylated at Ser317.
Recognizes the ~56 kDa Chk1 protein phosphorylated at Ser317 in UV-treated COS cells.
This PhosphoDetect Anti-Chk1 (pSer³¹⁷) Rabbit pAb is validated for use in Immunoblotting, Immunocytochemistry, Paraffin Sections for the detection of Chk1 (pSer³¹⁷).

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the Ser³¹⁷ phosphorylation site of human Chk1

Application


Immunoblotting (1:1000)
Immunocytochemistry (1:500, fluorescence)
Paraffin Sections (1:50)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM HEPES, 100 µg/ml BSA, 50% glycerol, pH 7.5.

Reconstitution

Do not aliquot.

Analysis Note

Positive Control
Lysates from COS cells treated with UV

Other Notes

Does not recognize unphosphorylated Chk1. Antibody should be titrated for optimal results in individual systems.
Zhao, H. and Piwnica-Worms, H. 2001. Mol. Cell. Biol.21, 4129.
Shieh, S.Y., et al. 2000. Genes Dev.14, 289.
Martinho, R.G., et al. 1998. EMBO J.17, 7239.
Zeng, Y., et al. 1998. Nature395, 507.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena Covelo-Molares et al.
Nucleic acids research, 49(19), 10895-10910 (2021-10-12)
N6-methyladenosine (m6A) and N6,2'-O-dimethyladenosine (m6Am) are two abundant modifications found in mRNAs and ncRNAs that can regulate multiple aspects of RNA biology. They function mainly by regulating interactions with specific RNA-binding proteins. Both modifications are linked to development, disease and
M Petroni et al.
Cell death and differentiation, 23(2), 197-206 (2015-06-13)
The MRE11/RAD50/NBS1 (MRN) complex is a major sensor of DNA double strand breaks, whose role in controlling faithful DNA replication and preventing replication stress is also emerging. Inactivation of the MRN complex invariably leads to developmental and/or degenerative neuronal defects
Yuki Funauchi et al.
Scientific reports, 5, 16497-16497 (2015-11-13)
Accumulation of iron in tissues increases the risk of cancer, but iron regulatory mechanisms in cancer tissues are largely unknown. Here, we report that p53 regulates iron metabolism through the transcriptional regulation of ISCU (iron-sulfur cluster assembly enzyme), which encodes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service