MilliporeSigma
  • The comprehensive interactomes of human adenosine RNA methyltransferases and demethylases reveal distinct functional and regulatory features.
The comprehensive interactomes of human adenosine RNA methyltransferases and demethylases reveal distinct functional and regulatory features.
Nucleic acids research (2021-10-12)
Helena Covelo-Molares, Ales Obrdlik, Ivana Poštulková, Michaela Dohnálková, Pavlína Gregorová, Ranjani Ganji, David Potěšil, Lisa Gawriyski, Markku Varjosalo, Štěpánka Vaňáčová
ABSTRACT
N6-methyladenosine (m6A) and N6,2'-O-dimethyladenosine (m6Am) are two abundant modifications found in mRNAs and ncRNAs that can regulate multiple aspects of RNA biology. They function mainly by regulating interactions with specific RNA-binding proteins. Both modifications are linked to development, disease and stress response. To date, three methyltransferases and two demethylases have been identified that modify adenosines in mammalian mRNAs. Here, we present a comprehensive analysis of the interactomes of these enzymes. PCIF1 protein network comprises mostly factors involved in nascent RNA synthesis by RNA polymerase II, whereas ALKBH5 is closely linked with most aspects of pre-mRNA processing and mRNA export to the cytoplasm. METTL16 resides in subcellular compartments co-inhabited by several other RNA modifiers and processing factors. FTO interactome positions this demethylase at a crossroad between RNA transcription, RNA processing and DNA replication and repair. Altogether, these enzymes share limited spatial interactomes, pointing to specific molecular mechanisms of their regulation.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
PhosphoDetect Anti-Chk1 (pSer³¹⁷) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, from mouse