Skip to Content
MilliporeSigma
All Photos(1)

Documents

CFUF01204

Milli-Q

Milli-Q® UF Ultrapure Cartridge Kit

An all-in-one consumable to remove trace organics, inorganic ions and bacteria

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

Quality Level

packaging

pkg of 1 kit

manufacturer/tradename

Milli-Q®

pore size

0.22 μm pore size

shipped in

ambient

General description

Milli-Q® UF Cartridge Kit is used with Milli-Q® UF Bowl-style water purification systems.

Features and Benefits

Includes one Super-C cartridge to eliminate dissolved organics, two nuclear-grade ion exchange cartridges to remove trace inorganic ions to 18 MΩ.cm resistivity, and one 0.22 μm Millipak® filter unit to remove bacteria.

Other Notes

Directions For Use:

  • Organism Retention: Microorganisms
  • Mode of Action: Filtering
  • Application: General laboratory analysis
  • Intended Use: Water purification
  • Instructions for Use: This item provides water filtered through 0.22 μm sterilizing-grade membranes. Refer to the system equipment user guide section "Using the System" (available on USB key in the system box).
  • Storage Statement: Store in a dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MILLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service