Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BI0223

Sigma-Aldrich

Capping Reagent A THF/Acetic Anhydride (9/1)

Novabiochem®

Synonym(s):

DNA Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

450 ML
$174.00
4 L
$653.00

$174.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
450 ML
$174.00
4 L
$653.00

About This Item

UNSPSC Code:
41116006

$174.00


Please contact Customer Service for Availability

Request a Bulk Order

product line

Novabiochem®

Quality Level

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

manufacturer/tradename

Novabiochem®

shipped in

ambient

storage temp.

room temp

General description

Biosynthesis Novabiochem®

Application

BI0223-0200 is packaged in 28/405 glass bottles
BI0223-0450 is packaged in 28/405 glass bottles
BI0223-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
Biosynthesis Novabiochem®

Analysis Note

Assay (acetic anhydride) (by density): 9.0-11.0% v/v
Assay (tetrahydrofuran) (by density): 89.0-91.0% v/v
Color (APHA): 10 max
Form: Clear liquid
Identity (IR-spectrum): Conforms
Water (H2O): 300ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service