Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AGX805

Sigma-Aldrich

2019-nCoV Nucleocapsid Recombinant Antigen,full length (Ag-5)

recombinant, expressed in E. coli, suitable for ELISA

Synonym(s):

SARS-CoV, SARS-CoV-2

Sign Into View Organizational & Contract Pricing

Select a Size

AGX805
$1,340.00

$1,340.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
AGX805
$1,340.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$1,340.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

form

liquid

technique(s)

ELISA: suitable

UniProt accession no.

shipped in

dry ice

Gene Information

SARS coronavirus ... N(43740575)

General description

This E. coli recombinant protein, Cat. No. AGX805 is tested for use in ELISA.

Specificity

SARS-CoV-2

Target description

Coronavirus disease 2019 (COVID-19) is an infectious disease caused by the virus strain severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), a virus closely related to the SARS virus. A SARS-CoV-2 nucleocapsid antigen can be used to support the detection of SARS-CoV-2 (COVID-19, 2019-nCoV).

Physical form

Purified
Column Purification
Purified protein in 25mM Tris-HCl with no preservatives, pH 8.0.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service