KK4752
Roche
KAPA PROBE FAST One-Step
Universal
Synonym(s):
RT-qPCR, probe qPCR, qPCR
Select a Size
$408.00
Select a Size
About This Item
$408.00
Recommended Products
Quality Level
usage
sufficient for 500 reactions
feature
dNTPs included: no
hotstart: no
packaging
pkg of 500 RXNS
manufacturer/tradename
Roche
technique(s)
RT-qPCR: suitable
input
purified DNA
detection method
probe-based
storage temp.
−20°C
General description
Application
- Gene expression analysis
- SNP/mutation analysis
- Gene knockdown validation
- RT-PCR[1]
Features and Benefits
- The kit is suitable for all fluorogenic probe-based technologies, including hybridization probes (e.g., FRET), hydrolysis probes (e.g., TaqMan) and displacement probes (e.g., molecular beacons).
- Use only gene-specific primers for one-step qRT-PCR.
- Optimal cDNA synthesis is achieved at 42°C for 5 min.
- 3 min at 95°C is sufficient for RT inactivation and DNA polymerase activation.
- Do not exceed 25 μL reaction volumes
Packaging
Quality
Preparation Note
Other Notes
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service