Skip to Content
MilliporeSigma
All Photos(3)

Documents

ABS981

Sigma-Aldrich

Anti-phospho-AMPK alpha-1 Antibody, (Thr479)

from rabbit, purified by affinity chromatography

Synonym(s):

5′-AMP-activated protein kinase catalytic subunit alpha-1, Thr479-phosphorylated, ACACA kinase, Thr479-phosphorylated, Acetyl-CoA carboxylase kinase, Thr479-phosphorylated, AMPK subunit alpha-1, Thr479-phosphorylated, AMPKa1, Thr479-phosphorylated, HMGCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rat (based on 100% sequence homology), porcine (based on 100% sequence homology), horse (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

dot blot: suitable
western blot: suitable

GenBank accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pThr479)

Gene Information

human ... PRKAA1(5562)

General description

5′-AMP-activated protein kinase catalytic subunit alpha-1 (EC 2.7.11.1; EC 2.7.11.26; EC 2.7.11.27; EC 2.7.11.31; UniProt Q13131; also known as ACACA kinase, Acetyl-CoA carboxylase kinase, AMPK subunit alpha-1, AMPKa1, HMGCR kinase, Hydroxymethylglutaryl-CoA reductase kinase, Tau-protein kinase PRKAA1) is encoded by the PRKAA1 (also known as AMPK1) gene (Gene ID 5562) in human. The yeast sucrose non-fermenting 1 (SNF1) and the AMP-activated protein kinase (AMPK) in drosphila and mammals are orthologous heterotrimeric serine-threonine kinases, each consisting of a catalytic α subunit and regulatory β and γ subunits, that sense decreased cellular energy levels when the AMP to ATP ratio rises due to a high rate of metabolism or decreased nutrient availability. AMP binding to the γ subunit results in an enhanced phosphorylation of the catalytic α subunit on an activation loop threonine and AMPK allosteric activation. Activated AMPK phosphorylates downstream targets, leading to energy conservation and inhibition of ATP consumption. For example, the high rate of ATP use in contracting muscle causes AMPK-mediated phosphorylation/inactivation of acetyl-CoA carboxylase (ACC), thereby increasing fatty acid oxidation and restoring cellular energy. AMPK functions as a positive regulator of autophagy by phosphorylating the protein kinase ULK1 that initiates autophagy as well as by inhibiting the mammalian TOR (mTOR) pathway activity via phosphorylating mTOR-binding partner raptor. In mammals, there exist two different α (encoded by the PRKAA1 and PRKAA2 gene), two different β (encoded by the PRKAB1 and PRKAB2 gene), and three different γ (encoded by the PRKAG1, PRKAG2, and PRKAG3 gene) subunits.

Specificity

The Thr479 numbering is based on the rat AMPK alpha-1 sequence shown in Fig 2A of Suzuki, T., et al. (2013). Mol. Cell. 50(3):407-419, which was cited for as Thr479 phosphorylation in subsequent publications (Coughlan, K.A., et al. (2015). PLoS One. 10(5):e0127388; Hawley, S.A., et al. (2014). Biochem. J. 459(2):275-287). The target threonine corresponds to Thr490 of the human/mouse/rat AMPK alpha-1 sequences published by UniProt (Q13131-1/Q5EG47/P54645) or Thr505 of human spliced isoform 2 (UniProt.Q13131-2). The target site sequence is not well conserved between AMPK alpha-1 (-SGTA(pT)PQR-) and AMPK alpha-2 (-SGSS(pT)PQR-).

Immunogen

KLH-conjugated linear peptide corresponding to the AMPK alpha-1 phosphorylation site sequence containing the phosphorylated target threonine.

Application

Detect AMPK threonine 479 phosphorylation using this rabbit polyclonal Anti-phospho-AMPK alpha-1 (Thr479), Cat. No. ABS981, validated for use in Dot Blot and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A 1:500 dilution from a representative lot detected AMPK alpha-1 phosphorylation induction upon 30-min serum stimulation of 6-hr serum-starved HEK293 cells (Courtesy of Bruce Kemp, Ph.D., St. Vincent′s Institute of Medical Research in Melbourne).

Western Blotting Analysis: A 1:1000 dilution from a representative lot detected GSK3beta-catalyzed phosphorylation of E. coli expressed, catalytically inactive AMPK alpha-1 recombinant protein (Courtesy of Bruce Kemp, Ph.D., St. Vincent′s Institute of Medical Research in Melbourne).

Quality

Evaluated by Peptide Dot Blot.

Dot Blot Analysis: A 1:1,000 dilution of this antibody detected 20-0.156 ng of immunogen peptide, but not the corresponding non-phosphorylated peptide.

Target description

~63 kDa observed. 64.01/65.52 kDa (human isoform 1/2) and 63.93/63.97 kDa (mouse/rat) calculate. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service