Skip to Content
MilliporeSigma
All Photos(2)

Documents

ABS453

Sigma-Aldrich

Anti-CHD7 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Chromodomain helicase DNA binding protein 7, CHD-7, IS3, KAL5, ATP-dependent helicase CHD7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CHD7(55636)

General description

Chromodomain-helicase-DNA-binding protein 7 (CHD7), also known as ATP-dependent helicase CHD7, belongs to a superfamily of proteins of the ATP-dependent chromatin remodeling enzymes that have a unique combination of functional domains, including two N-terminal chromodomains, a SNF2-like ATPase/helicase domain and a DNA-binding domain. CHD7 may play an important role in early embryonic development by affecting chromatin structure and gene expression. CHD7 interacts with embryonic stem (ES) cell master regulators OCT4/POU5F1, SOX2, and NANOG and is thought to modulate ES-specific gene transcription. Together with SOX2, CHD7 has been suggested to also regulate several human disease genes. Defects in CHD7 are suggested to be the cause of CHARGE syndrome (CHARGES) and idiopathic scoliosis type 3 (IS3). Mutations in CHD7 may also result in hypogonadotropic hypogonadism (HH5). CHD7 is widely expressed in fetal and adult tissues.

Immunogen

KLH-conjugated linear peptide corresponding to human CHD7.

Application

Anti-CHD7 Antibody is a highly specific rabbit polyclonal antibody, that targets CHD7 & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected CHD7 in mouse brain tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in SK-N-SH cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected CHD7 in 15 µg of SK-N-SH cell lysate.

Target description

~290 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 2 isoforms at ~128 kDa and ~336 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
SK-N-SH cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service