Skip to Content
MilliporeSigma
All Photos(3)

Documents

07-330-AF488

Sigma-Aldrich

Anti-Partitioning-defective 3 Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

Partitioning defective 3 homolog, PAR-3, PARD-3, Atypical PKC isotype-specific-interacting protein, ASIP, CTCL tumor antigen se2-5, PAR3-alpha, Partitioning-defective 3, Alexa Fluor 488 Conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

monkey, canine, human, rat, frog, mouse

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PARD3(56288)

General description

Partitioning defective 3 homolog (UniProt Q8TEW0; also known as ASIP, Atypical PKC isotype-specific-interacting protein, Bazooka, CTCL tumor antigen se2-5, Par-3 family cell polarity regulator alpha, Par-3 partitioning defective 3 homolog, PAR3-alpha, Protein phosphatase 1 regulatory subunit 118) is encoded by the PARD3 (also known as Baz, PAR3, PAR3A, PARD3A, PPP1R118) gene (Gene ID 56288) in human. Par3 is a PDZ (PSD95/Dlg/ZO-1) domain-containing membrane scaffold adapter protein that plays an important role in epithelial cell tight junction (TJ) formation and apico-basal polarity maintenance. Together with PAR6, atypical protein kinase C (aPKC), and the Rho GTPase CDC42, Par3 forms a complex that regulates cytoskeleton re-organization, post-golgi vesicle trafficking, microtubule network polarization, and asymmetric cell division. PAR6 interacts directly with aPKC isoforms iota/lambda and zeta to form the complex core. Recruitment of GTP-bound CDC42 to the PAR6-aPKC core induces aPKC phosphorylation/activation by PAR6, and subsequent PAR3 binding. Another quaternary apical junctional complex (AJC), Willin–Par6-aPKC–Par3, has also been reported. In this case, the complex-associated aPKC functions to phosphorylate ROCK, resulting in ROCK dissociation from the AJC and thereby preventing excessive ROCK-dependent apical constriction.

Specificity

Expected to react with all 11 spliced isofoms (UniProt Q8TEW0-1 to Q8TEW0-11).

Immunogen

GST fusion protein corresponding to residues 457-744 of mouse PAR-3 (accession number AY02657).

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Partitioning-defective 3 in A431, and NIH/3T3 cells.
The unconjugated antibody (Cat. No. 07-330) is shown to be suitable also for Western blotting, immunocytochemistry, and immunohistochemistry applications.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Partitioning-defective 3 Antibody, Alexa Fluor 488 Conjugate is validated for use in Immunocytochemistry for the detection of Partitioning-defective 3.

Quality

Evaluated by Immunocytochemistry in HUVECs.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Partitioning-defective 3 in HUVECs.

Target description

~180, 150, 100 kDa observed

Physical form

Protein A purified
Purified rabbit conjugate antibody in PBS with 15 mg/ml BSA and 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service