Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS2211

Sigma-Aldrich

Anti-USP16

Synonym(s):

Ubiquitin carboxyl-terminal hydrolase 16;EC:3.4.19.12;Deubiquitinating enzyme 16;Ubiquitin thioesterase 16;Ubiquitin-processing protease UBP-M;Ubiquitin-specific-processing protease 16

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$200.00

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$200.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

N/A, polyclonal

mol wt

calculated mol wt 93.57 kDa
observed mol wt ~105 kDa

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry (frozen sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... USP16(10600)

General description

Ubiquitin carboxyl-terminal hydrolase 16 (UniProt: Q9Y5T5; also known as EC:3.4.19.12, Deubiquitinating enzyme 16, Ubiquitin thioesterase 16, Ubiquitin-processing protease UBP-M, Ubiquitin-specific-processing protease 16) is encoded by the USP16 (also known as MSTP039) gene (Gene ID: 10600) in human. USP16 specifically deubiquitinates lysine 120 of histone H2A (H2AK119Ub), a specific tag for epigenetic transcriptional repression, thereby acting as a coactivator. Deubiquitination of histone H2A is reported to be a prerequisite for subsequent phosphorylation at serine 11 of histone H3 (H3S10ph) and is required for chromosome segregation when cells enter into mitosis. In resting B- and T-lymphocytes, phosphorylation by Aurora kinase B leads to enhancement in its deubiquitinase activity, thereby maintaining transcription in resting lymphocytes. USP16 also regulates Hox gene expression via histone H2A deubiquitination. It is not shown to deubiquitinate histone H2B. USP16 can contribute to somatic stem cell defects observed in Down syndrome. USP16 is triplicated in Down syndrome and its overexpression contributes to proliferation defects in stem cells. Reduction of USP16 levels results in increased proliferation capacity of Down syndrome fibroblasts. (Ref.: Joo, HY., et al. (2007). Nature. 449(7165);1068-1072).

Specificity

This rabbit polyclonal antibody detects Ubiquitin carboxyl-terminal hydrolase 16 (USP16). It targets an epitope within the N-terminal half.

Immunogen

GST-tagged recombinant fragment corresponding to the first 257 amino acids from the N-terminal half of human Ubiquitin carboxyl-terminal hydrolase 16 (USP16).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected UBP16 in HeLa cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected UBP16 in lysates from wild-type HEK293T cells, but not in lysate from HEK293T cells with USP16 knockout.

ELISA Analysis: A representative lot detected UBP16 in ELISA applications. (Yang, W., et. al. (2014). Nat Commun. 5:3818).

Immunohistochemistry Applications: A representative lot detected USP16 in Immunohistochemistry applications. (Yang, W., et. al. (2014). Nat Commun. 5:3818).

Flow Cytometry Analysis: A representative lot detected UBP16 in Flow Cytometry applications (Gu, Y., et. al. (2016). Proc. Natl. Acad. Sci. USA. 113(1); E51-60).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-USP16, Cat. No. ABS2211, is a rabbit polyclonal antibody that detects USP16 and is tested for use in ELISA, Flow Cytometry, Immunohistochemistry, and Western Blotting.

Physical form

Rabbit polyclonal antibody in serum with 0.05% sodium azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service