Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS2108

Sigma-Aldrich

Anti-HMG-Lysine

from rabbit

Synonym(s):

3-hydroxy-3-methyl-glutary-Lysine

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$231.00
100 μL
$590.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$231.00
100 μL
$590.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG

target post-translational modification

unmodified

General description

Acetylation of lysine residues in proteins has emerged as an important mechanism in regulation and coordination of normal and pathophysiological cell metabolism. Lysine HMGylation is a newer post-translational protein modification that occurs under normal basal conditions. In murine models of HMG-CoA lyase deficiency, higher levels of protein lysine HMGylation has been observed. This rabbit polyclonal antibody specifically detects chemically modified proteins containing hydroxymethylglutaryl-lysine modifications, but not unmodified proteins. (Ref.: Wagner, GR et al. (2017). Cell Metabolism 25(4); 823-837).

Specificity

This rabbit polyclonal antibody detects BSA-conjugated 3-hydroxy-3-methyl-glutaryl (HMG)-Lysine. It recognizes BSA chemically modified to contain HMG-lysine modifications, but not unmodified BSA.

Immunogen

3-hydroxy-3-methyl-glutaryl (HMG)-Lysine conjugated to BSA.
Epitope: unknown

Application

Anti-HMG-Lysine, Cat. No. ABS2108, is a rabbit polyclonal antibody that detects hydroxymethylglutaryl-lysine conjugated proteins and has been tested for use in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected HMG-Lysine in Western Blotting applications (Wagner, G.R., et. al. (2017). Cell Metab. 25(4):823-837; Anderson, K.A., et. al. (2017). Cell Metab. 25(4):838-855.e15).

Western Blotting Analysis: A 1:500 dilution from a representative lot detected HMG-Lysine in HMG-BSA conjugate (Courtesy of Dr Kristin Anderson from Duke University).

Quality

Evaluated by Western Blotting with HMG-BSA conjugate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected HMG-Lysine in HMG-Lysine-BSA conjugate.

Target description

~75 kDa observed (BSA conjugate). Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS with 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alec G Trub et al.
Nature communications, 13(1), 2542-2542 (2022-05-11)
Statins are a class of drug widely prescribed for the prevention of cardiovascular disease, with pleiotropic cellular effects. Statins inhibit HMG-CoA reductase (HMGCR), which converts the metabolite HMG-CoA into mevalonate. Recent discoveries have shown HMG-CoA is a reactive metabolite that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service