Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAK538

Sigma-Aldrich

Ammonia Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

detection method

colorimetric
fluorometric

relevant disease(s)

gastrointestinal diseases

shipped in

wet ice

storage temp.

−20°C

Related Categories

General description

Ammonia (NH3) or its ion form ammonium (NH4 +) is an important source of nitrogen for living systems. It is synthesized through amino acid metabolism and is toxic when present at high concentrations. In the liver, ammonia is converted to urea through the urea cycle. Elevated levels of ammonia in the blood (hyperammonemia) have been found in liver dysfunction (cirrhosis), while hypoammonemia has been associated with defects in the urea cycle enzymes (e.g. ornithine transcarbamylase).

Features and Benefits

  • Linear detection range: 24 to 1000 µM

Suitability

The kit is suitable for ammonia determination in biological samples (e.g. serum, plasma, urine, saliva, cell culture, etc).

Principle

The Ammonia Assay Kit is designed to directly measure NH3 and NH4 +. In this assay, NADH is converted to NAD+ in the presence of NH3, ketoglutarate and glutamate dehydrogenase. The decrease in optical density at 340 nm or fluorescence intensity at λEx = 360 nm/λEm = 450 nm is directly proportionate to the NH3 concentration in the sample.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service