Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

ABN427

Sigma-Aldrich

Anti-RalDH3 (ALDH1A3)

from rabbit, purified by affinity chromatography

Synonym(s):

Aldehyde dehydrogenase family 1 member A3, Aldehyde dehydrogenase 6, RALDH-3, RalDH3, Retinaldehyde dehydrogenase 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$622.00

$622.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$622.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$622.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

technique(s)

immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ALDH1A3(220)

General description

Aldehyde dehydrogenase family 1 member A3 (EC 1.2.1.5; UniProt Q9JHW9; also known as Aldehyde dehydrogenase 6, RALDH-3, RalDH3, Retinaldehyde dehydrogenase 3) is encoded by the Aldh1a3 (also known as Aldh6, Raldh3) gene (Gene ID 56847) in murine species. All-trans-retinoic acid (atRA) is a small molecule metabolite of retinol, also known as vitamine A, that plays an important role in the growth and development of many major organ systems, including the postnatal eye growth. The three retinaldehyde dehydrogenases, RALDH1-3 or ALDH1 1-3, catalyze the irreversible oxidation of retinaldehyde to form atRA. While retinaldehyde is formed via the reversible oxidation of retinol catalyzed by retinol dehydrogenases (RDHs). The three RALDH enzymes cooperatively mediate atRA signaling during eye development through tightly regulated spatiotemporal expression patterns. In the postnatal human eye, RALDH catalytic activity can be detected in the choroid, retinal pigment epithelium (RPE), and retina. RALDH2 is most abundant in the choroid and RPE, moderate abundance in the retina, and in relatively low abundance in sclera. RALDH1 is most abundant in the choroid, in moderate abundance in the sclera, and substantially reduced in the retina and RPE. RALDH3 is most abundantly expressed during embryonic eye development, but not detectable in adult ocular fundal tissues.

Immunogen

KLH-conjugated linear peptide corresponding to an internal sequence within the N-terminal half of mouse RalDH3.

Application

Anti-RalDH3 (ALDH1A3), Cat. No. ABN427, is a highly specific rabbit polyclonal antibody that targets RalDH3/ALDH1A3 and has been tested in Immunofluorescence and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A 1:500 dilutioin from a representative lot detected RalDH3 in 10 µg of human retina tissue lysate.

Immunofluorescence Analysis: A 1:150 dilutioin from a representative lot immunostained trabecula analge of the developing eye in paraffin-embedded embryonic E15 Sprague Dawley (SD) rat head sections by fluorescent immunohistochemistry (Courtesy of Anna Ashton, University of Aberdeen).

Western Blotting Analysis: A 1:1,500 dilutioin from a representative lot detected RalDH3 in 50 µg of new born (postnatal P0) Sprague Dawley (SD) rat whole eye tissue lysate, but not in adult SD rat brain lateral cortex and pons tissue lysates (Courtesy of Anna Ashton, University of Aberdeen).

Quality

Evaluated by Western Blotting in mouse retina tissue lysate.

Western Blotting Analysis: A 1:500 dilutioin of this antibody detected RalDH3 in 10 µg of mouse retina tissue lysate.

Target description

~53 kDa observed. 55.98/56.03/56.04 kDa (human/mouse/rat) calculated (Met1 removed).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oraly Sanchez-Ferras et al.
Nature communications, 12(1), 2627-2627 (2021-05-13)
The kidney and upper urinary tract develop through reciprocal interactions between the ureteric bud and the surrounding mesenchyme. Ureteric bud branching forms the arborized collecting duct system of the kidney, while ureteric tips promote nephron formation from dedicated progenitor cells.
Thabat Khatib et al.
Journal of Alzheimer's disease : JAD, 73(3), 935-954 (2019-12-31)
Retinoic acid has been previously proposed in the treatment of Alzheimer's disease (AD). Here, five transgenic mouse models expressing AD and frontotemporal dementia risk genes (i.e., PLB2APP, PLB2TAU, PLB1Double, PLB1Triple, and PLB4) were used to investigate if consistent alterations exist
Mutsuko Minata et al.
Cell reports, 26(7), 1893-1905 (2019-02-14)
Unresectable glioblastoma (GBM) cells in the invading tumor edge can act as seeds for recurrence. The molecular and phenotypic properties of these cells remain elusive. Here, we report that the invading edge and tumor core have two distinct types of
Kazuya Ono et al.
Nature communications, 11(1), 63-63 (2020-01-04)
Each vestibular sensory epithelium in the inner ear is divided morphologically and physiologically into two zones, called the striola and extrastriola in otolith organ maculae, and the central and peripheral zones in semicircular canal cristae. We found that formation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service