MilliporeSigma
  • Retinoic acid degradation shapes zonal development of vestibular organs and sensitivity to transient linear accelerations.
Retinoic acid degradation shapes zonal development of vestibular organs and sensitivity to transient linear accelerations.
Nature communications (2020-01-04)
Kazuya Ono, James Keller, Omar López Ramírez, Antonia González Garrido, Omid A Zobeiri, Hui Ho Vanessa Chang, Sarath Vijayakumar, Andrianna Ayiotis, Gregg Duester, Charles C Della Santina, Sherri M Jones, Kathleen E Cullen, Ruth Anne Eatock, Doris K Wu
ABSTRACT
Each vestibular sensory epithelium in the inner ear is divided morphologically and physiologically into two zones, called the striola and extrastriola in otolith organ maculae, and the central and peripheral zones in semicircular canal cristae. We found that formation of striolar/central zones during embryogenesis requires Cytochrome P450 26b1 (Cyp26b1)-mediated degradation of retinoic acid (RA). In Cyp26b1 conditional knockout mice, formation of striolar/central zones is compromised, such that they resemble extrastriolar/peripheral zones in multiple features. Mutants have deficient vestibular evoked potential (VsEP) responses to jerk stimuli, head tremor and deficits in balance beam tests that are consistent with abnormal vestibular input, but normal vestibulo-ocular reflexes and apparently normal motor performance during swimming. Thus, degradation of RA during embryogenesis is required for formation of highly specialized regions of the vestibular sensory epithelia with specific functions in detecting head motions.
Sigma-Aldrich
Sulforhodamine 101, Dye content ~95 %
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-RalDH3 (ALDH1A3), from rabbit, purified by affinity chromatography