Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABN1038

Sigma-Aldrich

Anti-Fbxo7 Antibody

Synonym(s):

F-box only protein 7|Fbxo7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

General description

F-box only protein 7 (UniProt Q9Y3I1) is encoded by the FBXO7 (Also known as FBX, FBX7, PKPS, FBX07, PARK15) gene in human (Gene ID 25793). The Skp1-Cul1-F box protein (SCF) E3 ubiquitin ligase complexes rely on their F-box protein component for substrate recognition. In addition to Fbxs, to which Fbxo7 belongs, there exist two other classes of F-box proteins, Fbws and Fbls. Fbxo7 recruits substrate via its C-terminal proline rich domain and associates with the adapter protein Skp1 via its F-box domain. In addition to mediating SCF-dependent ubiquitination, Fbxo7 is also involved in SCF-independent activities. Fbxo7 is reported to participate in autophagic clearance of damaged mitochondria (mitophagy) in response to mitochondrial depolarisation through direct interaction with PINK1 (PARK6) and Parkin (PARK2), while Parkinson′s disease-associated Fbxo7 mutations affect Parkin mitochondrial translocation and Parkin-mediated mitophagy.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human Fbxo7.

Application

Anti-Fbxo7 Antibody is an antibody against Fbxo7 for use in Western Blotting, Immunohistochemistry, Immunocytochemistry.
Immunohistochemistry Analysis: A 1:400 dilution from a representative lot detected Fbxo7 in human tonsil tissue.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Fbxo7 using U2OS cells transfected with constructs expres

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1.0 ug/mL of this antibody detected Fbxo7 in 10 ug of Jurkat cell lysate.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service