Skip to Content
MilliporeSigma
All Photos(2)

Documents

ABE2605

Sigma-Aldrich

Anti-H3K4me3Q5histaminyl

Synonym(s):

Histone H3 trimethylated Lysine 4 Histaminylated glutamine5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 15.51 kDa

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

dot blot: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

trimethylation (Lys4), histaminylation (Gln5)

Gene Information

human ... HIST3H3(8290)

General description

Histone H3.1t (UniProt: Q16695; also known as H3/t, H3t, H3/g) is encoded by the HIST3H3 (also known as H3FT) gene (Gene ID: 8290) in human. Histones are highly conserved basic nuclear proteins that are responsible for the nucleosome structure of chromatin in eukaryotes. They play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which DNA is wrapped in repeating units, called nucleosomes, which limits DNA accessibility to the cellular machineries that require DNA as a template. Histone H3 features a main globular domain and a long N-terminal tail, which protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. The amino-terminal tails of histone proteins are subject to posttranslational modifications, including acetylation and methylation, which recruit downstream regulatory factors, influence chromatin structure, and are critical determinants of transcription. Acetylation of histone H3 occurs at several different lysine positions in the histone tail and is performed by a family histone acetyltransferases. Acetylation is generally associated with transcriptional activity and methylation of lysine and arginine residues can either activate or repress depending on the residue modified. There is a significant correlation between acetylation of histones H3 in promoter regions and transcriptional activity. The presence of H3K4me3 has been correlated with transcriptionally active promoters of genes and its levels positively correlate with gene expression levels. H3K4me3 has been implicated in a number of nuclear processes, including Pol II-mediated transcription, pre-mRNA splicing, DNA recombination, and DNA repair. Histaminylation of glutamine 5 on histone H3 (H3Q5Histaminyl) is recently identified as a permissive post-translational modification that coexists with adjacent lysine 4 trimethylation. (Ref.: Zhao, S., et al. (2021). Proc. Natl. Acad. Sci. USA. 118(6):e2016742118; Diehl , KL., and Muir, TW. (2020). Nat. Chem. Biol. 16; 620-629; Bernstein, BE., et al. (2002). Proc. Natl. Acad. Sci. USA. 99(13);8695-8700).

Specificity

This rabbit polyclonal antibody specifically detects Histone H3.1 with trimethylated lysine 4 and histaminylated glutamine 5.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids from the N-terminal region of human Histone H3.1 with trimethylated lysine 4 and histaminylated glutamine 5.

Application

Quality Control Testing

Evaluated by DOT Blot analysis with K3K4me3Q5Histaminyl peptide.

Dot Blot Analysis: A 1:1,000 dilution of this antibody detected H3K4me3Q5Histaminyl peptide, but not the unmodified H3Q5 peptide.

Tested Applications


Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected Histone H3.1 trimethylated on lysine 4 and histaminylated on glutamine 5 (H3K4me3Q5Histaminyl) in RN46A, KU812, SK-N-SH & HEK293T whole cell lysates. (Data courtesy of Dr. Ian Maze s Lab, Icahn School of Medicine at Mount Sinai, New York).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-H3K4me3Q5histaminyl, Cat. No. ABE2605, is a rabbit polyclonal antibody that detects Histone H3.1 trimethylated on lysine 4 and histaminylated on glutamine 5 and is tested for use in Dot Blot and Western Blotting.

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service