Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB4200621

Sigma-Aldrich

Monoclonal Anti-NG2/CSPG4 antibody produced in mouse

clone 2164B6, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2164B6, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: 2.5-5 μg/test using SK-Mel-28 cells.
immunofluorescence: 2.5-5 μg/mL using A375 cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSPG4(1464)

General description

Nerve/glial antigen 2 (NG2), a high molecular mass melanoma- associated antigen (HMW-MAA), is a large integral membrane, cell surface type I transmembrane protein that is covalently modified with chondroitin sulfate glycosaminoglycan.

Immunogen

a recombinant extracellular portion of human NG2 proteoglycan

Application

Monoclonal Anti-NG2/CSPG4 antibody produced in mouse used in flow cytometry and immunocytochemistry.

Biochem/physiol Actions

Nerve/glial antigen 2 (NG2) plays a central role in linking multiple oncogenic pathways required for malignant progression. NG2 expression in radial growth phase (RGP) human melanomas enables migration, protease activation and epithelial to mesenchymal transition, which is important in primary tumors for facilitating progression from a radial to vertical growth phase phenotype. It regulates sustained, high-level activation of key survival and growth pathways, integrin-regulated focal adhesion kinase (FAK), extracellular signal-regulated kinases 1 and 2 (ERK 1,2) and phosphoinositide 3-kinase (PI3K)/AKT pathways.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan S Lord et al.
The FEBS journal, 280(10), 2490-2510 (2013-02-20)
Proteoglycans are ubiquitous dynamic molecules that are made up of a protein core to which specific linear glycosylation structures, known as glycosaminoglycans, have been covalently coupled. They have roles in many biological and pathological processes, which have been shown to
Matthew A Price et al.
Pigment cell & melanoma research, 24(6), 1148-1157 (2011-10-19)
Chondroitin sulfate proteoglycan 4 (CSPG4), a transmembrane proteoglycan originally identified as a highly immunogenic tumor antigen on the surface of melanoma cells, is associated with melanoma tumor formation and poor prognosis in certain melanomas and several other tumor types. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service