Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB2910

Sigma-Aldrich

Anti-Mcl-1 Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MCL1(4170)

Specificity

Recognizes Human Mcl-1. Not reactive with other members of the Bcl-2 family, as verified by immunoblot.

Immunogen

Synthetic peptide corresponding to amino acids 121-139 of human Mcl-1.

Application

Immunoblot 1:2,000

Immunoprecipitation. Precipitates a 37-39 kDa protein, which appears as a doublet in some cells.

Immunohistochemistry (formalin-fixed, paraffin-embedded tissues)

The following tissues/cells can serve as positive controls: tonsil, tonsillar B cells, B-cell lines Reh, Daudi, and Karpas 422.

Optimal working dilutions for AB2910 must be determined by end user.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
This Anti-Mcl-1 Antibody is validated for use in IP, WB, IH(P) for the detection of Mcl-1.

Physical form

Liquid antiserum.

Storage and Stability

Store at -20°C for up to 12 months in undiluted aliquots. Avoid repeated freeze-thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yixiang Li et al.
Theranostics, 10(17), 7656-7670 (2020-07-21)
Prostate cancer is the second leading cause of cancer-related death in the United States. As a first line treatment for hormone-refractory prostate cancer, docetaxel (DTX) treatment leads to suboptimal effect since almost all patients eventually develop DTX resistance. In this
Nicole M Smith et al.
Journal of neuroinflammation, 15(1), 201-201 (2018-07-10)
Following injury to the central nervous system, increased microglia, secretion of pro- and anti-inflammatory cytokines, and altered blood-brain barrier permeability, a hallmark of degeneration, are observed at and immediately adjacent to the injury site. However, few studies investigate how regions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service