Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB15510

Sigma-Aldrich

Anti-Neuroligin 2 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NLGN2(57555)

General description

Neuroligins are Type I membrane proteins enriched in synaptic plasma membranes and clustered in synaptic clefts and postsynaptic densities. They have been characterized as neuronal cell surface proteins and are thought to be involved in cell-cell-interactions by forming intercellular junctions through binding to beta-neurexins. They play a major role in the formation or maintenance of synaptic junctions. They are also thought to be involved in the specification of excitatory synapses. Neuroligins interact with beta-neurexins and this interaction is involved in the formation of functional synapses.

Specificity

Neuroligin 2 (NLGN2).

Immunogen

Synthetic peptide near the C-terminus of human Neuroligin 2

Application

Anti-Neuroligin 2 Antibody is an antibody against Neuroligin 2 for use in WB, ELISA.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western blot: 1-10 μg/mL using ECL.
Immunohistochemistry: Not tested. It is recommended that the antibody be tried at 2-20 μg/mL
ELISA: 1:10,000-1:100,000 using 50-100 ng of control peptide/well.
Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Liquid in PBS with 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service