Skip to Content
MilliporeSigma
All Photos(1)

Documents

228180

Sigma-Aldrich

Cholesterol Esterase, Microbial

Cholesterol Esterase, Microbial, CAS 9026-00-0, is a native enzyme that catalyzes hydrolysis and formation of cholesterol esters, ranging from cholesterol acetate to cholesterol stearate.

Synonym(s):

Cholesterol Esterase, Microbial, CE, Sterolester Acylhydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

Quality Level

form

powder

specific activity

≥100 units/mg protein
≥70 units/mg dry wt.

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

light brown

solubility

10 mM potassium phosphate/sodium hydroxide, 0.1% BSA, pH 7.5: soluble

foreign activity

Catalase ≤1.5%

shipped in

ambient

storage temp.

−20°C

General description

Native, microbial cholesterol esterase. Catalyzes hydrolysis and formation of cholesterol esters, ranging from cholesterol acetate to cholesterol stearate.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol cholesterol ester per min at 37°C, pH 6.5.

Reconstitution

Reconstitute just prior to use. Enzyme will lose activity in solution upon storage.

Other Notes

Allain, C.C., et al. 1974. Clin. Chem.20, 470.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service