Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.55090

Sigma-Aldrich

Oxime resin HL (100-200 mesh)

Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$163.00
5 G
$650.00

$163.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$163.00
5 G
$650.00

About This Item

UNSPSC Code:
13111023
NACRES:
NA.22

$163.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

oxime

storage temp.

2-30°C

General description

Nucleophile cleavable resin for synthesis of C-terminal modified peptides by Boc SPPS

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Linkage

Replaces: 01-64-0294

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 1.0 - 1.5 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene - 1% DVB) 100 - 200 mesh.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Anhydrous HF is the preferred reagent for peptide cleavage from Boc-based resins, versatile and effective for various peptide synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service