Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.55021

Sigma-Aldrich

4-Sulfamylbutyryl AM resin

Novabiochem®

Synonym(s):

4-Sulfamylbutyryl AM Solid Support Resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111023
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

sulfonamide

storage temp.

2-30°C

General description

Safety-catch resin for synthesis of C-terminally modified peptides by Fmoc SPPS. Linker is activated to cleavage by nucleophiles by methylation using TMS-diazomethane.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Application

  • The First Solid-phase Synthesis and Structural Studies on Phakellistatin 15: This study describes the use of 4-sulfamylbutyryl AM resin for the solid-phase synthesis of Phakellistatin 15, highlighting the resin′s role in peptide synthesis and its effectiveness in loading Fmoc-Leu-OH. (Shaheen et al., 2016).

Linkage

Replaces: 01-64-0152

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.60 - 1.20 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene-1 % DVB), 200 -400 mesh.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service