Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.52034

Sigma-Aldrich

Fmoc-Arg(Pmc)-OH

Novabiochem®

Synonym(s):

Fmoc-Arg(Pmc)-OH, N-α-Fmoc-N G-(2,2,5,7,8-pentamethylchroman-6-sulfonyl)-L-arginine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$149.00
25 G
$585.00
100 G
$1,750.00
500 G
$8,360.00
1 KG
$15,200.00

$149.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$149.00
25 G
$585.00
100 G
$1,750.00
500 G
$8,360.00
1 KG
$15,200.00

About This Item

Empirical Formula (Hill Notation):
C35H42N4O7S
CAS Number:
Molecular Weight:
662.80
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

$149.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥85.0% (acidimetric)
≥98% (TLC)
≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

70-120 °C (decomposition)

application(s)

peptide synthesis

functional group

amine

storage temp.

15-25°C

SMILES string

[S](=O)(=O)(\N=C(\NCCC[C@H](NC(=O)OCC3c4c(cccc4)c5c3cccc5)C(=O)O)/N)c1c(c2c(c(c1C)C)OC(CC2)(C)C)C

InChI

1S/C35H42N4O7S/c1-20-21(2)31(22(3)23-16-17-35(4,5)46-30(20)23)47(43,44)39-33(36)37-18-10-15-29(32(40)41)38-34(42)45-19-28-26-13-8-6-11-24(26)25-12-7-9-14-27(25)28/h6-9,11-14,28-29H,10,15-19H2,1-5H3,(H,38,42)(H,40,41)(H3,36,37,39)/t29-/m0/s1

InChI key

QTWZCODKTSUZJN-LJAQVGFWSA-N

General description

An excellent derivative for the introduction of Arg in Fmoc SPPS. The Pmc group is removed by TFA in 1-3 hours [1,2,3]. For discussions on the side-reactions associated with the use of sulfonyl-based arginine protection, see [4,5,6,7,8,9,10,11]. In the synthesis of peptides containing both Arg and Trp, it is recommended that this derivative is used in conjunction with Fmoc-Trp(Boc)-OH (852050) [5,6,11].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS


Literature references

[1] R. Ramage, et al. (1987) Tetrahedron Lett., 28, 2287.
[2] J. Green, et al. (1988) Tetrahedron Lett., 29, 4341.
[3] R. Ramage, et al. (1991) Tetrahedron Lett., 47, 6353.
[4] P. Sieber (1987) Tetrahedron Lett., 28, 1637.
[5] C. G. Fields, et al. (1993) Tetrahedron Lett., 34, 6661.
[6] H. Choi, et al. (1993) Int. J. Peptide Protein Res., 42, 58.
[7] A. Stierandova, et al. (1994) Int. J. Peptide Protein Res., 43, 31.
[8] P. M. Fischer, et al. (1992) Int. J. Peptide Protein Res., 40, 19.
[9] S. A. G. Beck, et al. (1991) Int. J. Peptide Protein Res., 38, 25.
[10] E. Jaeger, et al. (1993) Biol. Chem. Hoppe-Seyler, 374, 349.
[11] P. White in ′Peptides, Chemistry & Biology, Proc. 12th American Peptide Symposium′, J. A. Smith & J. E. Rivier (Eds), ESCOM, Leiden, 1992, pp. 537.

Linkage

Replaces: 04-12-1073

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011C)): ≥ 98 %
Purity (TLC(157B)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (12,5 mmol in 25 ml DMF): clearly soluble
Assay (acidimetric): ≥ 85.0 %
Water (K. F.): ≤ 1.50 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service