Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.22260

Sigma-Aldrich

Succinic acid

for synthesis

Synonym(s):

Butanedioic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$44.20
1 KG
$93.00
50 KG
$2,880.00

$44.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$44.20
1 KG
$93.00
50 KG
$2,880.00

About This Item

Linear Formula:
HOOCCH2CH2COOH
CAS Number:
Molecular Weight:
118.09
Beilstein:
1754069
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-740-4
NACRES:
NA.22

$44.20


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥99% (acidimetric)

form

solid

autoignition temp.

470 °C

potency

2260 mg/kg LD50, oral (Rat)

pH

2.7 (20 °C, 10 g/L in H2O)

bp

235 °C (lit.)
235 °C/1013 hPa

mp

184-186 °C (lit.)

transition temp

flash point 206 °C

solubility

water: 58 g/L at 20 °C

density

1.56 g/cm3 at 20 °C

bulk density

940 kg/m3

storage temp.

2-30°C

SMILES string

OC(=O)CCC(O)=O

InChI

1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)

InChI key

KDYFGRWQOYBRFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 184 °C
Melting range (upper value): ≤ 189 °C
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service