Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20448

Sigma-Aldrich

2,5-Dichlorophenol

for synthesis

Synonym(s):

2,5-Dichlorophenol, 2,5-Dichlo-1-hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,5-(Cl)2C6H3OH
CAS Number:
Molecular Weight:
163.00
MDL number:
UNSPSC Code:
12352104
EC Index Number:
209-520-4

Quality Level

Assay

≥99.0% (GC)

form

crystals

potency

580 mg/kg LD50, oral (Rat)

bp

211 °C/1013 hPa

mp

56-58 °C

solubility

2000 mg/L (experimental)

storage temp.

2-30°C

InChI

1S/C6H4Cl2O/c7-4-1-2-5(8)6(9)3-4/h1-3,9H

InChI key

RANCECPPZPIPNO-UHFFFAOYSA-N

Application

2,5-Dichlorophenol for synthesis. CAS 583-78-8, chemical formula 2,5-(Cl)₂C₆H₃OH.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 53 °C
Melting range (upper value): ≤ 57 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service