Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.40083

Sigma-Aldrich

(2-Dimethylaminoethyl) methacrylate

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

(2-Dimethylaminoethyl) methacrylate, 2-Propenoic acid-2-methyl-(2-dimethylamino)ethylester

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$44.70
250 ML
$71.40
1 L
$198.00

$44.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$44.70
250 ML
$71.40
1 L
$198.00

About This Item

Empirical Formula (Hill Notation):
C8H15NO2
CAS Number:
Molecular Weight:
157.21
MDL number:
UNSPSC Code:
12352108
EC Index Number:
220-688-8
NACRES:
NA.22

$44.70


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

255 °C

potency

1751 mg/kg LD50, oral (Rat)

expl. lim.

1.20 % (v/v)

bp

182-192 °C/1013 hPa

mp

-50 °C

transition temp

flash point 65 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

N(CCOC(=O)C(=C)C)(C)C

InChI

1S/C8H15NO2/c1-7(2)8(10)11-6-5-9(3)4/h1,5-6H2,2-4H3

InChI key

JKNCOURZONDCGV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.930 - 0.934
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service