Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20287

Sigma-Aldrich

Chlorocyclohexane

for synthesis

Synonym(s):

Chlorocyclohexane, Cyclohexyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$50.80
1 L
$133.00

$50.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$50.80
1 L
$133.00

About This Item

Empirical Formula (Hill Notation):
C6H11Cl
CAS Number:
Molecular Weight:
118.60
MDL number:
UNSPSC Code:
12352101
EC Index Number:
208-806-6
NACRES:
NA.22

$50.80


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

7 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

290 °C

potency

3000 mg/kg LD50, oral (Rat)

expl. lim.

1.1-7.5 % (v/v)

bp

141-143 °C/1013 hPa

mp

-44 °C

transition temp

flash point 34 °C

density

1.00 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC1CCCCC1

InChI

1S/C6H11Cl/c7-6-4-2-1-3-5-6/h6H,1-5H2

InChI key

UNFUYWDGSFDHCW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.996 - 0.999
Identity (IR): passes test

Pictograms

FlameEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F

Flash Point(C)

34 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service