8.20051
Aluminium
(granulated) for synthesis
Synonym(s):
Aluminium
Select a Size
$36.20
Select a Size
About This Item
$36.20
Recommended Products
Quality Level
bp
2467 °C/1013 hPa
mp
660 °C
density
2.70 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
[Al]
InChI
1S/Al
InChI key
XAGFODPZIPBFFR-UHFFFAOYSA-N
Analysis Note
Identity (ICP-OES): passes test
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service