Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14588

Sigma-Aldrich

1,2-Propylene glycol diacetate

for synthesis

Synonym(s):

1,2-Propylene glycol diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O4
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352108
EC Index Number:
210-817-6
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

431 °C

potency

13530 mg/kg LD50, oral (Rat)

expl. lim.

2.8-12.7 % (v/v)

bp

190-191 °C/1013 hPa

transition temp

flash point 87 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H12O4/c1-5(11-7(3)9)4-10-6(2)8/h5H,4H2,1-3H3

InChI key

MLHOXUWWKVQEJB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.056 - 1.058
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service