Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.21014

Sigma-Aldrich

[2-(2-Butoxyethoxy)ethyl] acetate

for synthesis

Synonym(s):

[2-(2-Butoxyethoxy)ethyl] acetate, Butyl diglycol acetate, Acetic acid 2-(2-butoxyethoxy)ethyl ester, Diethylene glycol monobutyl ether acetate, Diethylene glycol butyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O4
CAS Number:
Molecular Weight:
204.26
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-685-9
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
244-250 °C/1013 hPa
vapor pressure:
<0.01 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

<0.01 hPa ( 20 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

290 °C

potency

6500 mg/kg LD50, oral (Rat)
5640 mg/kg LD50, skin (Rabbit)

expl. lim.

1.0-5.3 % (v/v)

dilution

(for synthesis)

pH

3-4 (20 °C, 50 g/L in H2O)

bp

244-250 °C/1013 hPa

mp

-32 °C

transition temp

flash point 102 °C

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCOCCCC)CCOC(=O)C

InChI

1S/C10H20O4/c1-3-4-5-12-6-7-13-8-9-14-10(2)11/h3-9H2,1-2H3

InChI key

VXQBJTKSVGFQOL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.976 - 0.978
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service