Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.08096

Sigma-Aldrich

Tetrabromophthalic anhydride

for synthesis

Synonym(s):

Tetrabromophthalic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$86.10

$86.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$86.10

About This Item

Empirical Formula (Hill Notation):
C8Br4O3
CAS Number:
Molecular Weight:
463.70
MDL number:
UNSPSC Code:
12352115
EC Index Number:
211-185-4
NACRES:
NA.22

$86.10


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

14.67 Pa ( 20 °C)

Quality Level

form

powder

potency

>10000 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

mp

271-274 °C

density

2.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Brc1c2c(c(c(c1Br)Br)Br)C(=O)OC2=O

InChI

1S/C8Br4O3/c9-3-1-2(8(14)15-7(1)13)4(10)6(12)5(3)11

InChI key

QHWKHLYUUZGSCW-UHFFFAOYSA-N

Analysis Note

Assay (ex Br): ≥ 98.0 %
Melting range (lower value): ≥ 270 °C
Melting range (upper value): ≤ 280 °C
Identity (IR): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service