Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

174010

Supelco

Halogen lamp module

for use with SPECTROQUANT® Prove 100

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$2,540.00

$2,540.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
$2,540.00

About This Item

UNSPSC Code:
41116116
NACRES:
SG.01

$2,540.00


Please contact Customer Service for Availability

Request a Bulk Order

compatibility

for use with SPECTROQUANT® Prove 100

shipped in

ambient

storage temp.

no temp limit

General description

Prove 100 Spectroquant®

Application

  • Incoherent broadband cavity-enhanced absorption spectroscopy for sensitive measurement of nutrients and microalgae.: This research explores the use of halogen lamp modules in incoherent broadband cavity-enhanced absorption spectroscopy, demonstrating their utility in the sensitive detection of nutrients and microalgae. The study underscores the importance of these light sources in enhancing the sensitivity and accuracy of analytical measurements (Zhang et al., 2022).
  • Fluorescence remission spectroscopy of psoriatic lesions and the effect of topical anthralin therapy.: This article investigates the application of halogen lamp modules in fluorescence remission spectroscopy for examining psoriatic lesions and evaluating the effectiveness of topical anthralin therapy. The results suggest significant applications in medical and dermatological research, providing insights into treatment efficacy and skin condition monitoring (Wollina et al., 2009).

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service