Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00588

Sigma-Aldrich

2-Hydroxyethyl methacrylate

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

2-Hydroxyethyl methacrylate, 2-Hydroxyethyl 2-methylpropenoate, HEMA, Methacrylic acid 2-hydroxyethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$67.70
2.5 L
$172.00
25 KG
$1,630.00

$67.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$67.70
2.5 L
$172.00
25 KG
$1,630.00

About This Item

Linear Formula:
CH2=C(CH3)COOCH2CH2OH
CAS Number:
Molecular Weight:
130.14
MDL number:
UNSPSC Code:
12352108
EC Index Number:
212-782-2
NACRES:
NA.22

$67.70


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1.3 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

potency

5050 mg/kg LD50, oral (Rat)
>3000 mg/kg LD50, skin (Rabbit)

bp

250 °C/1013 hPa

mp

-60 °C

transition temp

flash point 101 °C

density

1.07 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCO)C(=O)C(=C)C

InChI

1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3

InChI key

WOBHKFSMXKNTIM-UHFFFAOYSA-N

Application

  • Preparation and application of naringenin imprinted cryogel membranes for selectively separation of naringenin from natural environment: This study focuses on using 2-hydroxyethyl methacrylate as a co-monomer in cryogels designed for specific molecule adsorption from natural sources (E Demirtaş, M Odabaşı - 2024, Springer).
  • Metallization of polypropylene substrates through surface functionalization and physical vapor deposition of chromium coatings: Investigates the enhancement of adhesive properties on polypropylene substrates using 2-hydroxyethyl methacrylate, advancing material engineering and surface treatments (N Fumagalli et al., 2024, Elsevier).

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.069 - 1.072
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service