Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72350

Sigma-Aldrich

BacMagic-3 DNA Kit - Novagen

Synonym(s):

BacMagic DNA, BacMagic DNA Kits

Sign Into View Organizational & Contract Pricing

Select a Size

5 REACTIONS
$1,140.00

$1,140.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 REACTIONS
$1,140.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

$1,140.00


Please contact Customer Service for Availability

Request a Bulk Order

manufacturer/tradename

Novagen®

Quality Level

availability

not available in Japan

storage condition

OK to freeze

shipped in

wet ice

storage temp.

2-8°C

General description

The BacMagic-3 DNA Kit provides a positive selection for creating baculovirus recombinants in preparation for recombinant protein expression in insect cells. The kit improves on the traditional method for generating recombinant baculoviruses by eliminating the tedious, time-consuming steps of plaque purification. Derived from an AcNPV genome, BacMagic-3 DNA is deleted for a portion of the essential open reading frame (ORF) 1629, preventing nonrecombinant virus form replicating in insect cells. The nonessential chitinase (chiA) gene is deleted to improve production of secreted and membrane target proteins. Additionally, the v-cath gene, a cathespsin-like cysteine protease, is deleted resulting in significantly improved quality and yield for most target proteins. Unlike BacMagic-2, the BacMagic-3 genome is deleted for an additional three viral genes (p10, p24, and p26). These genes are nonessential to virus in tissue culture and improve protein expression. The BacMagic-3 DNA Kit contains the BacMagic-3 DNA, Insect GeneJuice® Transfection Reagent, and a positive control plasmid to monitor co-transfection efficiency.

Components

  • 0.5 µg- BacMagic-3 DNA
  • 50 µl- Insect GeneJuice Transfection Reagent
  • 2 µg- Transfection Control Plasmid

Legal Information

GENEJUICE is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service