Skip to Content
MilliporeSigma
All Photos(1)

Documents

71557

Sigma-Aldrich

pIEx-10 DNA - Novagen®

Novagen′s pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera derived insect cells.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106612
NACRES:
NA.51

Quality Level

form

solid

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

General description

The pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells. Transcription is driven by the AcNPV-derived hr5 enhancer and immediate early promoter, IE1. The pIEx-10 contains the coding sequence for the mouse IgM signal sequence to allow secretion of the expressed protein. It also encodes an N-terminal 8 aa Strep•Tag II coding sequence followed by an enterokinase protease cleavage site and a multiple cloning site, with the option to include a C-terminal His•Tag coding sequence with 10 histidines. Additionally, pIEx-10 is available as an Ek/LIC vector. The Ek/LIC vector is prepared for rapid directional cloning of PCR-amplified DNA. Using specifically designed primers for amplification, inserts can be efficiently cloned without the need for restriction enzyme digestion or ligation.

Warning

Toxicity: Irritant (B)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service