Skip to Content
MilliporeSigma
All Photos(1)

Documents

71553-M

Sigma-Aldrich

pET-51b(+) DNA - Novagen

Novagen′s pET-51b(+) vector is designed for cloning and high-level expression of target proteins fused with the 8 aa Strep-Tag II coding sequence that is cleavable with enterokinase (Ek) protease.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106602
NACRES:
NA.51

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

−70°C

General description

The pET-51b(+) vector is designed for cloning and high-level expression of target proteins fused with the 8 aa Strep•Tag II coding sequence that is cleavable with enterokinase (Ek) protease. The plasmid contains a strong T7lac promoter, an optimized RBS, the coding sequence for the Ek protease cleavage site (AspAspAspAspLys), and a multiple cloning site that contains restriction enzyme sites found in many other Novagen expression vectors to facilitate insert transfer. An optional C-terminal His•Tag coding sequence with 10 histidines is compatible with purification, detection, and quantification.

The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock). Please contact technical service if you need additional information.




Commercial use of this Product requires a commercial license from EMD Millipore Corporation. Commercial use shall include but not be limited to (1) use of the Product or its components in manufacturing; (2) use of the Product or its components to provide a service, information, or data to others in exchange for consideration; (3) use of the Product or its components (or any derivatives thereof) for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the Product or its components, whether or not such Product or its components are resold for use in research. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of this Product.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service