Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71530-M

Sigma-Aldrich

T7 Tail Fiber Monoclonal Antibody

Novagen®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

antibody product type

primary antibodies

manufacturer/tradename

Novagen®

storage condition

OK to freeze

isotype

IgG2a

storage temp.

−20°C

target post-translational modification

unmodified

General description

Specific immunocapture of T7 bacteriophage
The T7 Tail Fiber Monoclonal Antibody is a mouse monoclonal antibody (IgG2a) directed against the tail-fiber protein of the T7 bacteriophage. The antibody can be used to immunoprecipitate T7 phage or T7 tail-fiber from solution using an optimized immunoprecipitation GrabIt-based protocol (Protein G Plus/Protein A agarose). Just 2.5 µg of antibody is sufficient to immunoprecipitate at least 1 × 1010 T7 phage. The antibody is supplied at 1 mg/ml and is suitable for use with the T7Select Phage Display System and the immunoprecipitation-based GrabIt Protein G Plus/Protein A Kit. Not suitable forimmunoblotting.

Warning

Toxicity: Highly Toxic (H)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service