Skip to Content
MilliporeSigma

1.37126

SAFC

Hexylene glycol

EMPROVE® EXPERT, NF

Pharma Manufacturing

Synonym(s):

1,1,3-Trimethyltrimethylenediol, 2,4-Dihydroxy-2-methylpentane, 2-Methyl-2,4-pentanediol, 2-Methylpentane-2,4-diol, Diolane, Isol, (±)-2-Methyl-2,4-pentanediol, MPD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2C(CH3)2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1098298
MDL number:
UNSPSC Code:
12161700
EC Index Number:
203-489-0

grade

GMP

Quality Level

Agency

NF

vapor density

4.1 (vs air)

vapor pressure

0.02 mmHg ( 20 °C)

product line

EMPROVE® EXPERT

form

liquid

expl. lim.

7.4 %

impurities

low bacterial content

refractive index

n20/D 1.427 (lit.)

bp

197 °C (lit.)

mp

−40 °C (lit.)

density

0.925 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical

storage temp.

2-25°C

SMILES string

CC(O)CC(C)(C)O

InChI

1S/C6H14O2/c1-5(7)4-6(2,3)8/h5,7-8H,4H2,1-3H3

InChI key

SVTBMSDMJJWYQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right critical raw material that matches your quality expectation can be challenging.
Especially if you are advancing through clinical phases, changing regulatory requirements canraise several questions.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Hexylene glycol (or the racemic 2-methylpentane-2,4-diol) is used as a critical raw material in several industries and applications. For pharmaceutical applications, our Emprove® Expert NF grade product can be used in formulation as an emulsifier, or as a critical process chemical in API synthesis.

Packaging

1.37126.1000 // 1 liter // Glass bottle

1.37126.5000 // 5 liter // Stainless steel drum

1.37126.9020 // 20 liter // Stainless steel drum

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service