Skip to Content
MilliporeSigma
All Photos(1)

Documents

71412

Millipore

Chicken Egg White Lysozyme Solution, 10 mg/ml

A ready-to-use, stabilized lysozyme solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

biological source

chicken egg white

Quality Level

form

solution

manufacturer/tradename

Novagen®

storage condition

OK to freeze

concentration

10 mg/mL

technique(s)

DNA purification: suitable
protein purification: suitable

suitability

suitable for nucleic acid purification

application(s)

life science and biopharma

shipped in

wet ice

storage temp.

−20°C

General description

Chicken Egg White Lysozyme Solution is a ready-to-use, stabilized solution of lysozyme. The enzyme catalyses the hydrolysis of N-acetylmuramide linkages in bacterial cell walls. The solution can be used for the purification of both DNA and protein. We recommend Chicken Egg White Lysozyme Solution for Gram-positive bacterial lysis. The solution is supplied as 10 mg/ml lysozyme in 25 mM sodium acetate with 50% glycerol, pH 4.5 (≥250 KU/ml). The product is stable at -20°C.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit of Chicken Egg White Lysozyme is defined as the amount of enzyme that causes a decrease of 0.001 A₄₅₀ unit per minute at 25°C, pH 6.2, using a suspension of Micrococcus lysodeikticus as the substrate.

Physical form

10 mg/ml lysozyme in 25 mM sodium acetate pH 4.5 with 50% glycerol.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service