Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693015-2

Supelco

Pressure Dispenser, 1/4" x 1/4"

for use with NOWPAK®

Synonym(s):

hplc solvents

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$3,730.00

$3,730.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
$3,730.00

About This Item

UNSPSC Code:
41105107

$3,730.00


Please contact Customer Service for Availability

Request a Bulk Order

compatibility

for use with NOWPAK®

shipped in

ambient

General description

for NOWPak II, NOWPak®

Legal Information

NOWPAK is a registered trademark of Advanced Technology Materials, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service