Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6.17990

Sigma-Aldrich

Brij 35

Synonym(s):

Brij® 35

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)nC12H26O
CAS Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

SMILES string

O(CCO)CCCCCCCCCCCC

InChI

1S/C14H30O2/c1-2-3-4-5-6-7-8-9-10-11-13-16-14-12-15/h15H,2-14H2,1H3

InChI key

SFNALCNOMXIBKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>300.2 °F

Flash Point(C)

> 149 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service