Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

614005

Sigma-Aldrich

Tobramycin, Free Base

Aminoglycoside antibiotic active against Gram negative bacteria. Inhibits myeloperoxidase-dependent oxidant cell injury.

Synonym(s):

Tobramycin, Free Base

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H37N5O9
CAS Number:
Molecular Weight:
467.51
MDL number:
UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white to off-white

solubility

water: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C18H37N5O9/c19-3-9-8(25)2-7(22)17(29-9)31-15-5(20)1-6(21)16(14(15)28)32-18-13(27)11(23)12(26)10(4-24)30-18/h5-18,24-28H,1-4,19-23H2/t5-,6+,7+,8-,9+,10+,11-,12+,13+,14-,15+,16-,17+,18+/m0/s1

InChI key

NLVFBUXFDBBNBW-PBSUHMDJSA-N

General description

Aminoglycoside antibiotic active against Gram negative bacteria. Inhibits myeloperoxidase-dependent oxidant cell injury.
Aminoglycoside antibiotic. Inhibits myeloperoxidase-dependent oxidant cell injury.

Biochem/physiol Actions

Primary Target
myeloperoxidase-dependent oxidant cell injury

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Other Notes

Cantin, A. and Woods, D.E. 1993. J. Clin. Invest. 91, 38.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service