Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BP333

Tobramycin

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Nebramycin Factor 6, O-[3-Amino-3-deoxy-α-D-glucopyranosyl-(1→6)]-O-[2,6-diamino-2,3,6-trideoxy-α-D-ribohexopyranosyl-(1→4)]-2-deoxy-D-streptamine

Sign Into View Organizational & Contract Pricing

Select a Size

350 MG
$325.00

$325.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
350 MG
$325.00

About This Item

Empirical Formula (Hill Notation):
C18H37N5O9
CAS Number:
Molecular Weight:
467.51
Beilstein:
1357507
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$325.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

tobramycin

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@@H](N)[C@H]3O)[C@H]2O)[C@H](N)C[C@@H]1O

InChI

1S/C18H37N5O9/c19-3-9-8(25)2-7(22)17(29-9)31-15-5(20)1-6(21)16(14(15)28)32-18-13(27)11(23)12(26)10(4-24)30-18/h5-18,24-28H,1-4,19-23H2/t5-,6+,7+,8-,9+,10+,11+,12+,13+,14-,15+,16-,17+,18+/m0/s1

InChI key

NLVFBUXFDBBNBW-SNGYORCQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support

Application

Tobramycin BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Tobramycin and Dexamethasone Eye Drops, Suspension
  • Apramycin Premix
  • Tobramycin Inhalation Powder, Hard Capsule
  • Apramycin Sulfate
  • Tobramycin Nebuliser Solution
  • Tobramycin Oral Solution
  • Tobramycin Eye Drops
  • Tobramycin Injection
  • Tobramycin Sulfate
  • Apramycin Veterinary Oral Powder
  • Biochem/physiol Actions

    Tobramycin is an aminoglycoside.
    Mode of Action: Binds to 70S ribosomal subunit; inhibits translocation; elicits miscoding.
    Spectrum of Activity: Gram negative bacteria. Not effective against Enterococci.

    Packaging

    Unit quantity: 350 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Health hazard

    Signal Word

    Warning

    Hazard Statements

    Hazard Classifications

    Repr. 2

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service