Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

566332

Sigma-Aldrich

Sirt1 Inhibitor VII, Inauhzin

The Sirt1 Inhibitor VII, Inauhzin controls the biological activity of Sirt1. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Sirt1 Inhibitor VII, Inauhzin, 10-[2-(5H-[1,2,4]triazino[5,6-b]indol-3-ylthio)butanoyl]-10H-phenothiazine, INZ

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$217.00

$217.00


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
$217.00

About This Item

Empirical Formula (Hill Notation):
C25H19N5OS2
Molecular Weight:
469.58
UNSPSC Code:
12352200
NACRES:
NA.28

$217.00


Please contact Customer Service for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

potency

0.7 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable phenothiazine derivative that selectively inhibits SirT1 (IC50 ≤2.0 µM), but not SirT2, SirT3, or HCAC8 (IC50 >50 µM), activity. Effectively elevates cellular p53 lysine acetylation (Effective conc. 2 µM), thereby protecting p53 lysine residues from MDM2-mediated ubiquitination without inhibiting MDM2 activity toward deacetylated p53. Shown to inhibit cancer cell growth in a p53-dependent manner (IC50 = 2.0 and 15.7µM, respectively, against HCT116p53+/+ and HCT116p53-/- cells) and synergize with Nutlin-3 (Cat. Nos. 444143, https://www.sigmaaldrich.com/US/en/product/mm/444151" target="_blank"">444151, & 444152) in blocking HCT116p53+/+ proliferation in vitro (1 µM INZ & 4 µM Nutlin-3) and inducing HCT116p53+/+ apoptosis in mice in vivo (15 mg INZ/kg/d i.p; 150 mg Nut-3/kg/12 h p.o.).

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Zhang, Q., et al. 2012. EMBO Mol. Med.4, 298.
Zhang, Y., et al. 2012. Cancer Biol. Ther.13, 915

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service